Assay Details
| Gene Symbol: Atrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172774.3 | NP_766362.2 | 4 - 5 | 786 | 79 | |
| GenBank mRNA | AK031257.1 | - | 4 - 5 | 776 | 79 | |
| AK171423.1 | - | 4 - 5 | 741 | 79 | ||
| BC078458.1 | - | 4 - 5 | 733 | 6832436 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atrip
Genomic Map