Assay Details
| Gene Symbol: Ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172144.3 | NP_742156.2 | 1 - 2 | 438 | 74 | |
| GenBank mRNA | AK035077.1 | - | 1 - 2 | 218 | 74 | |
| AK132175.1 | - | 1 - 2 | 439 | 74 | ||
| AK169510.1 | - | 1 - 2 | 371 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r3a
Genomic Map