Assay Details
| Gene Symbol: Ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161362.3 | NP_001154834.1 | 5 - 6 | 3077 | 69 | |
| NM_172144.3 | NP_742156.2 | 4 - 5 | 917 | 69 | ||
| GenBank mRNA | AK035077.1 | - | 4 - 5 | 697 | 69 | |
| AK076163.1 | - | 4 - 5 | 2129 | 69 | ||
| AK132175.1 | - | 4 - 5 | 918 | 69 | ||
| BC042841.1 | - | 4 - 5 | 2038 | 69 | ||
| BC042842.1 | - | 4 - 5 | 2511 | 69 | ||
| BC151170.1 | - | 4 - 5 | 2904 | 9088059 | 69 | |
| BC151171.1 | - | 4 - 5 | 2899 | 9088060 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r3a
Genomic Map