Assay Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198168.3 | NP_937811.1 | 5 - 6 | 1192 | 108 | |
| GenBank mRNA | AK034167.1 | - | 5 - 6 | 1211 | 108 | |
| AK155872.1 | - | 5 - 6 | 1185 | 108 | ||
| AK171041.1 | - | 5 - 6 | 1163 | 108 | ||
| AK171178.1 | - | 5 - 6 | 1193 | 108 | ||
| AK171430.1 | - | 5 - 6 | 1200 | 108 | ||
| BC058977.1 | - | 5 - 6 | 983 | 5687054 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5b
Genomic Map