Assay Details
| Gene Symbol: Ccm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145536.3 | NP_663511.2 | 3 - 4 | 336 | 132 | |
| GenBank mRNA | BC020535.1 | - | 3 - 4 | 336 | 4240594 | 132 |
More Information
Set Membership: |
|

Ccm2l
Genomic Map