Assay Details
| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033435.3 | NP_001028607.1 | 2 - 3 | 204 | 77 | |
| NM_001271372.1 | NP_001258301.1 | 2 - 3 | 204 | 77 | ||
| NM_001271373.1 | NP_001258302.1 | 2 - 3 | 204 | 77 | ||
| NM_001271374.1 | NP_001258303.1 | 2 - 3 | 204 | 77 | ||
| NM_001271375.1 | NP_001258304.1 | 2 - 3 | 204 | 77 | ||
| NR_073165.1 | 2 - 3 | 204 | 77 | |||
| GenBank mRNA | AB542953.1 | - | 2 - 3 | 162 | 77 | |
| AK136488.1 | - | 2 - 3 | 164 | 77 | ||
| AK137500.1 | - | 2 - 3 | 163 | 77 | ||
| AK151814.1 | - | 2 - 3 | 162 | 77 | ||
| AK153506.1 | - | 2 - 3 | 112 | 77 | ||
| AK169925.1 | - | 2 - 3 | 167 | 77 | ||
| AK170947.1 | - | 2 - 3 | 163 | 77 | ||
| AK171381.1 | - | 2 - 3 | 163 | 77 | ||
| BC139224.1 | - | 2 - 3 | 190 | 8862246 | 77 | |
| BC139225.1 | - | 2 - 3 | 190 | 8862247 | 77 | |
| BC145580.1 | - | 2 - 3 | 190 | 9054146 | 77 | |
| BC145582.1 | - | 2 - 3 | 190 | 9054148 | 77 | |
| BY197245.1 | - | 2 - 3 | 208 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK153506.1 transcripts.
Set Membership: |
|

Milr1
Genomic Map