Assay Details
| Gene Symbol: Prss45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153172.1 | NP_694812.1 | 1 - 2 | 142 | 75 | |
| GenBank mRNA | AF392062.1 | - | 1 - 2 | 142 | 75 | |
| AK077121.1 | - | 1 - 2 | 140 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss45
Genomic Map