Assay Details
| Gene Symbol: Socs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007707.3 | NP_031733.1 | 1 - 2 | 488 | 113 | |
| GenBank mRNA | AK047165.1 | - | 1 - 2 | 308 | 113 | |
| AK139241.1 | - | 1 - 2 | 309 | 113 | ||
| AK150719.1 | - | 1 - 2 | 308 | 113 | ||
| AK151761.1 | - | 1 - 2 | 308 | 113 | ||
| AK152441.1 | - | 1 - 2 | 308 | 113 | ||
| AK152468.1 | - | 1 - 2 | 308 | 113 | ||
| AK152514.1 | - | 1 - 2 | 308 | 113 | ||
| AK152686.1 | - | 1 - 2 | 308 | 113 | ||
| AK155097.1 | - | 1 - 2 | 272 | 113 | ||
| AK157708.1 | - | 1 - 2 | 294 | 113 | ||
| AK159395.1 | - | 1 - 2 | 306 | 113 | ||
| AK170406.1 | - | 1 - 2 | 306 | 113 | ||
| AK172399.1 | - | 1 - 2 | 308 | 113 | ||
| BC052031.1 | - | 1 - 2 | 284 | 5707539 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047165.1, AK139241.1, AK150719.1, AK151761.1, AK152441.1, AK152468.1, AK152514.1, AK152686.1, AK157708.1, AK159395.1, AK170406.1, AK172399.1, BC052031.1, NM_007707.3 transcripts.
Set Membership: |
|

Socs3
Genomic Map