Assay Details
| Gene Symbol: P2rx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008771.3 | NP_032797.3 | 4 - 5 | 594 | 66 | |
| GenBank mRNA | AK035304.1 | - | 4 - 5 | 595 | 66 | |
| BC015084.1 | - | 4 - 5 | 608 | 4189541 | 66 | |
| X84896.1 | - | 4 - 5 | 588 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

P2rx1
Genomic Map