Assay Details
| Gene Symbol: Ceacam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054059.1 | NP_473400.1 | 5 - 6 | 1485 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam3| Gene Symbol: Ceacam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054059.1 | NP_473400.1 | 5 - 6 | 1485 | 111 | |
Set Membership: |
|