Assay Details
| Gene Symbol: Ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198613.2 | NP_941015.2 | 1 - 2 | 94 | 88 | |
| GenBank mRNA | AK146584.1 | - | 1 - 2 | 95 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146584.1,NM_198613.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap2s1
Genomic Map