Assay Details
| Gene Symbol: Emx2os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY117414.1 | - | LIT1111 EMX2OS |
1 - 2 | 539 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | empty spiracles homeobox 2 | Emx2 | Pdo | Anti-sense | Chr. 19 - 59458116 - 59465357 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2700089I24 gene | 2700089I24Rik | 34.5kb | Anti-sense | Chr. 19 - 59493136 - 59559391 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Emx2os
Genomic Map