Assay Details
| Gene Symbol: Emx2os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002863.2 | 2 - 3 | 766 | 97 | |||
| GenBank mRNA | AK033011.1 | - | 2 - 3 | 415 | 97 | ||
| AY117414.1 | - | LIT1111 EMX2OS |
2 - 3 | 97 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ domain containing 8 | Pdzd8 |
A630041P07Rik, AV329468, AW228944, Pdzk8 |
-84.2kb | Sense | Chr. 19 - 59296084 - 59345780 on NCBI Build GRCm38 |
| Downstream | empty spiracles homeobox 2 | Emx2 | Pdo | 25.1kb | Anti-sense | Chr. 19 - 59458116 - 59465357 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Emx2os
Genomic Map