Assay Details
| Gene Symbol: Polr3gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293701.1 | NP_001280630.1 | 2 - 3 | 82 | ||
| NM_027241.4 | NP_081517.2 | 3 - 4 | 82 | |||
| GenBank mRNA | AK008069.1 | - | 3 - 4 | 364 | 82 | |
| BC027083.1 | - | 3 - 4 | 5357914 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr3gl
Genomic Map