Assay Details
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009358.3 | NP_033384.2 | 5 - 6 | 750 | 140 | |
| GenBank mRNA | AB053464.1 | - | 5 - 6 | 695 | 140 | |
| AF316616.2 | - | 5 - 6 | 607 | 140 | ||
| AK154050.1 | - | 5 - 6 | 140 | |||
| AK160587.1 | - | 5 - 6 | 691 | 140 | ||
| AY338234.1 | - | 5 - 6 | 680 | 140 | ||
| BC010716.1 | - | 5 - 6 | 663 | 2655861 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5d
Genomic Map