Assay Details
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009358.3 | NP_033384.2 | 9 - 10 | 1203 | 68 | |
| GenBank mRNA | AB053464.1 | - | 9 - 10 | 1148 | 68 | |
| AF316616.2 | - | 9 - 10 | 1060 | 68 | ||
| AK154050.1 | - | 9 - 10 | 1179 | 68 | ||
| AK160587.1 | - | 9 - 10 | 1144 | 68 | ||
| AY338234.1 | - | 9 - 10 | 1133 | 68 | ||
| BC010716.1 | - | 9 - 10 | 1116 | 2655861 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5d
Genomic Map