Assay Details
| Gene Symbol: Mea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277310.1 | NP_001264239.1 | 1 - 2 | 55 | 82 | |
| GenBank mRNA | BG085669.2 | - | 1 - 2 | 50 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277310.1 transcripts.
Set Membership: |
|

Mea1
Genomic Map