Assay Details
| Gene Symbol: Mea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277309.1 | NP_001264238.1 | 1 - 2 | 143 | 85 | |
| GenBank mRNA | BY707372.1 | - | 1 - 2 | 126 | 85 | |
| CJ042312.1 | - | 1 - 2 | 131 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mea1
Genomic Map