Assay Details
| Gene Symbol: Prss36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081374.1 | NP_001074843.2 | 11 - 12 | 2058 | 66 | |
| GenBank mRNA | BC044738.2 | - | 2 - 3 | 320 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss36
Genomic Map