Assay Details
| Gene Symbol: Mss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029104.1 | NP_083380.1 | 1 - 2 | 269 | 76 | |
| GenBank mRNA | BC119773.1 | - | 1 - 2 | 162 | 40107223 | 76 |
| BC119816.1 | - | 1 - 2 | 162 | 40107221 | 76 | |
| BY341628.1 | - | 1 - 2 | 271 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119773.1, BC119816.1, NM_029104.1 transcripts.
Set Membership: |
|

Mss51
Genomic Map