Assay Details
| Gene Symbol: Pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011057.3 | NP_035187.2 | 3 - 4 | 1063 | 88 | |
| GenBank mRNA | AF162784.1 | - | 3 - 4 | 253 | 88 | |
| BC023427.1 | - | 2 - 3 | 187 | 88 | ||
| BC053430.1 | - | 3 - 4 | 1063 | 6330609 | 88 | |
| BC064056.1 | - | 3 - 4 | 730 | 1380782 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfb
Genomic Map