Assay Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028792.3 | NP_083068.1 | 2 - 3 | 1274 | 66 | |
| GenBank mRNA | AK004913.1 | - | 2 - 3 | 1223 | 66 | |
| AK154799.1 | - | 2 - 3 | 1323 | 66 | ||
| BC006928.1 | - | 2 - 3 | 422 | 2655826 | 66 | |
| BC086769.1 | - | 2 - 3 | 394 | 5707990 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Josd1
Genomic Map