Assay Details
| Gene Symbol: Prss41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027644.1 | NP_081920.1 | 2 - 3 | 103 | 131 | |
| GenBank mRNA | AB049453.1 | - | 2 - 3 | 103 | 131 | |
| AK016509.1 | - | 2 - 3 | 81 | 108 | ||
| AK049297.1 | - | 2 - 3 | 120 | 131 | ||
| BC145586.1 | - | 2 - 3 | 119 | 131 | ||
| BC150797.1 | - | 2 - 3 | 119 | 9087708 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB049453.1, AK049297.1, BC150797.1, NM_027644.1 transcripts.
Set Membership: |
|

Prss41
Genomic Map