Assay Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011945.2 | NP_036075.2 | 7 - 8 | 1420 | 112 | |
| GenBank mRNA | AB014614.1 | - | 7 - 8 | 1397 | 112 | |
| AF117340.1 | - | 7 - 8 | 1420 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k1
Genomic Map