Assay Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011945.2 | NP_036075.2 | 9 - 10 | 1689 | 74 | |
| GenBank mRNA | AF117340.1 | - | 9 - 10 | 1689 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map3k1
Genomic Map