Assay Details
| Gene Symbol: Ppp1r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199149.3 | NP_954600.1 | 1 - 2 | 592 | 76 | |
| GenBank mRNA | AK050946.1 | - | 1 - 2 | 593 | 76 | |
| AK160935.1 | - | 1 - 2 | 591 | 76 | ||
| BC070402.1 | - | 1 - 2 | 440 | 5696223 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r37
Genomic Map