Assay Details
| Gene Symbol: Ppp1r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199149.3 | NP_954600.1 | 5 - 6 | 952 | 55 | |
| GenBank mRNA | AK050946.1 | - | 5 - 6 | 953 | 55 | |
| AK160935.1 | - | 5 - 6 | 953 | 55 | ||
| BC070402.1 | - | 5 - 6 | 800 | 5696223 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r37
Genomic Map