Assay Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178775.4 | NP_848890.3 | 12 - 13 | 2989 | 66 | |
| GenBank mRNA | BC058403.1 | - | 3 - 4 | 1688 | 6812243 | 66 |
| BG079911.2 | - | 2 - 3 | 363 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kc1
Genomic Map