Assay Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178775.4 | NP_848890.3 | 2 - 3 | 251 | 68 | |
| GenBank mRNA | AK030313.1 | - | 229 | 68 | ||
| AK149858.1 | - | 2 - 3 | 216 | 68 | ||
| BB619353.1 | - | 2 - 3 | 252 | 68 | ||
| CJ045347.1 | - | 2 - 3 | 252 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kc1
Genomic Map