Assay Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178775.4 | NP_848890.3 | 9 - 10 | 1199 | 83 | |
| GenBank mRNA | AK044809.1 | - | 6 - 7 | 918 | 83 | |
| AK049003.1 | - | 2 - 3 | 147 | 83 | ||
| AK149858.1 | - | 9 - 10 | 1164 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kc1
Genomic Map