Assay Details
| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008939.2 | NP_032965.1 | 7 - 8 | 1747 | 103 | |
| GenBank mRNA | AK132633.1 | - | 7 - 8 | 1536 | 103 | |
| AK186311.1 | - | 1 - 2 | 76 | 103 | ||
| BC031429.1 | - | 7 - 8 | 1649 | 3665834 | 103 | |
| D89871.1 | - | 7 - 8 | 1745 | 103 | ||
| Y13192.1 | - | 7 - 8 | 1524 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss12
Genomic Map