Assay Details
| Gene Symbol: Ppp2r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138748.5 | NP_620087.3 | 4 - 5 | 601 | 58 | |
| GenBank mRNA | AK031591.1 | - | 4 - 5 | 602 | 58 | |
| AK046288.1 | - | 4 - 5 | 405 | 58 | ||
| AK049184.1 | - | 4 - 5 | 592 | 58 | ||
| AK150029.1 | - | 4 - 5 | 596 | 58 | ||
| AK160980.1 | - | 4 - 5 | 456 | 58 | ||
| AK195195.1 | - | 1 - 2 | 55 | 58 | ||
| AY035997.1 | - | 4 - 5 | 454 | 58 | ||
| BC006626.1 | - | 4 - 5 | 517 | 3158086 | 58 | |
| BC052852.1 | - | 4 - 5 | 527 | 30058414 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r4
Genomic Map