Assay Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050359.1 | - | 9 - 10 | 1166 | 61 | |
| AK173334.1 | - | 6 - 7 | 2785 | 61 | ||
| BC085286.1 | - | 10 - 11 | 1292 | 61 | ||
| BC157930.1 | - | 17 - 18 | 2597 | 61 | ||
| BC172022.1 | - | 17 - 18 | 2599 | 61 | ||
| BC172146.1 | - | 37 - 38 | 5164 | 61 | ||
| DQ924536.1 | - | 37 - 38 | 5461 | 61 | ||
More Information
Set Membership: |
|

Ubr3
Genomic Map