Assay Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153587.2 | NP_705815.1 | 2 - 3 | 319 | 72 | |
| GenBank mRNA | AK030051.1 | - | 2 - 3 | 376 | 72 | |
| AK032316.1 | - | 2 - 3 | 412 | 72 | ||
| AK046319.1 | - | 2 - 3 | 96 | 72 | ||
| AK135786.1 | - | 227 | 72 | |||
| AY341873.1 | - | 2 - 3 | 308 | 72 | ||
| AY341881.1 | - | 2 - 3 | 308 | 72 | ||
| BC035298.1 | - | 2 - 3 | 319 | 4021865 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka5
Genomic Map