Assay Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153587.2 | NP_705815.1 | 10 - 11 | 1391 | 132 | |
| GenBank mRNA | AK030051.1 | - | 9 - 10 | 1332 | 132 | |
| AK032316.1 | - | 10 - 11 | 1484 | 132 | ||
| AK046319.1 | - | 10 - 11 | 1168 | 132 | ||
| AK160758.1 | - | 3 - 4 | 350 | 132 | ||
| AY341873.1 | - | 11 - 12 | 1380 | 132 | ||
| AY341881.1 | - | 10 - 11 | 1380 | 132 | ||
| BC035298.1 | - | 11 - 12 | 1391 | 4021865 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka5
Genomic Map