Assay Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146199.1 | NP_001139671.1 | 15 - 16 | 3100 | 70 | |
| NM_011877.2 | NP_036007.2 | 15 - 16 | 3235 | 70 | ||
| GenBank mRNA | BC049893.1 | - | 13 - 14 | 2982 | 70 | |
| BC055942.1 | - | 3 - 4 | 1624 | 70 | ||
| BC066856.1 | - | 13 - 14 | 1494 | 70 | ||
| BC098500.1 | - | 15 - 16 | 3100 | 30081068 | 70 | |
| D37801.1 | - | 15 - 16 | 3184 | 70 | ||
| D83072.1 | - | 2 - 3 | 496 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn21
Genomic Map