Assay Details
| Gene Symbol: Spint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016907.3 | NP_058603.2 | 6 - 7 | 1122 | 97 | |
| GenBank mRNA | AF099018.1 | - | 6 - 7 | 1077 | 97 | |
| AK017342.1 | - | 5 - 6 | 1004 | 97 | ||
| BC005769.1 | - | 5 - 6 | 1009 | 2650110 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spint1
Genomic Map