Assay Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032465.2 | NP_115854.2 | 7 - 8 | 1228 | 96 | |
| GenBank mRNA | AK040539.1 | - | 7 - 8 | 1232 | 96 | |
| AK154076.1 | - | 7 - 8 | 1198 | 96 | ||
| AK156470.1 | - | 7 - 8 | 1229 | 96 | ||
| AY029156.1 | - | 7 - 8 | 1089 | 96 | ||
| BC052865.1 | - | 7 - 8 | 1125 | 30072864 | 96 | |
More Information
Set Membership: |
|

Cd96
Genomic Map