Assay Details
| Gene Symbol: Polr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145632.2 | NP_663607.1 | 3 - 4 | 348 | 84 | |
| GenBank mRNA | AK146429.1 | - | 3 - 4 | 373 | 84 | |
| AK161617.1 | - | 3 - 4 | 369 | 84 | ||
| BC002306.1 | - | 3 - 4 | 348 | 3591968 | 84 | |
| BC092087.1 | - | 3 - 4 | 258 | 5042770 | 84 | |
| BC125612.1 | - | 3 - 4 | 332 | 40130127 | 84 | |
| BC125638.1 | - | 3 - 4 | 332 | 40130153 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Polr2h
Genomic Map