Assay Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159561.1 | NP_001153033.1 | 6 - 7 | 834 | 112 | |
| NM_011028.2 | NP_035158.2 | 7 - 8 | 915 | 112 | ||
| GenBank mRNA | AK138541.1 | - | 7 - 8 | 767 | 112 | |
| BC117713.2 | - | 6 - 7 | 696 | 40088045 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx6
Genomic Map