Assay Details
| Gene Symbol: Rpl7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025433.3 | NP_079709.2 | 6 - 7 | 779 | 155 | |
| GenBank mRNA | AK005264.1 | - | 6 - 7 | 768 | 155 | |
| AK007884.1 | - | 6 - 7 | 768 | 155 | ||
| AK008672.1 | - | 6 - 7 | 769 | 155 | ||
| AK090082.1 | - | 6 - 7 | 779 | 155 | ||
| AK152288.1 | - | 5 - 6 | 708 | 155 | ||
| AK166499.1 | - | 6 - 7 | 783 | 155 | ||
| AK167036.1 | - | 6 - 7 | 779 | 155 | ||
| BC003907.1 | - | 1 - 2 | 212 | 155 | ||
| BC030165.1 | - | 6 - 7 | 777 | 5027519 | 155 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl7l1
Genomic Map