Assay Details
| Gene Symbol: Gm38758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC062895.1 | - | 1 - 2 | 282 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NEDD4 binding protein 2-like 1 | N4bp2l1 |
2410024N18Rik, AI428195, B230342M21Rik |
-9.3kb | Sense | Chr. 5 - 150571643 - 150594525 on NCBI Build GRCm38 |
| Downstream | NEDD4 binding protein 2-like 2 | N4bp2l2 |
2700092H06Rik, zag1 |
29.0kb | Sense | Chr. 5 - 150635973 - 150665612 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm38758
Genomic Map