Assay Details
| Gene Symbol: Iscu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025526.4 | NP_079802.1 | 2 - 3 | 245 | 116 | |
| GenBank mRNA | AK009021.1 | - | 2 - 3 | 246 | 116 | |
| AK009421.1 | - | 2 - 3 | 249 | 116 | ||
| AK039834.1 | - | 2 - 3 | 287 | 116 | ||
| AK083247.1 | - | 2 - 3 | 253 | 116 | ||
| BC028800.1 | - | 2 - 3 | 223 | 116 | ||
| BC048409.1 | - | 2 - 3 | 242 | 6484953 | 116 | |
| BC096049.1 | - | 2 - 3 | 243 | 3466271 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Iscu
Genomic Map