Assay Details
| Gene Symbol: Iscu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025526.4 | NP_079802.1 | 4 - 5 | 433 | 124 | |
| GenBank mRNA | AK009021.1 | - | 4 - 5 | 434 | 124 | |
| AK009421.1 | - | 4 - 5 | 437 | 124 | ||
| AK039834.1 | - | 4 - 5 | 475 | 124 | ||
| AK083247.1 | - | 4 - 5 | 441 | 124 | ||
| BC028800.1 | - | 4 - 5 | 411 | 124 | ||
| BC048409.1 | - | 4 - 5 | 430 | 6484953 | 124 | |
| BC096049.1 | - | 4 - 5 | 431 | 3466271 | 124 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular iron ion homeostasis
iron-sulfur cluster assembly
[2Fe-2S] cluster assembly
protein maturation by iron-sulfur cluster transfer
iron ion binding
ferrous iron binding
protein complex scaffold
iron-sulfur transferase activity
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding
cellular iron ion homeostasis
iron-sulfur cluster assembly
[2Fe-2S] cluster assembly
protein maturation by iron-sulfur cluster transfer
iron ion binding
ferrous iron binding
protein complex scaffold
iron-sulfur transferase activity
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23999975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23999976 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020549 | AK157864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020552 | BC028800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020552 | BC048409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020552 | BC096049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020552 | AK009021.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020552 | AK009421.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317259 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113605 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020549 | AK157864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020552 | BC028800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020552 | BC048409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020552 | BC096049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020552 | AK009021.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020552 | AK009421.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24000006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23999977 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01191724 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113605 | - | - |
| Whole Mouse Genome G4122A | A_51_P316188 | BC028800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316188 | BC048409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316188 | BC096049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316188 | AK009021.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316188 | AK009421.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112365 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122076 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04103673 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23999975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23999976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24000000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23999994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04130439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04567357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04310805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04567349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04134583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04103676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04310802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23999995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24000019 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10524420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524421 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452579_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461108 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10524417 | - | - |
| Mouse Genome 430 2.0 Array | 1452579_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452579_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66383_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66383_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500010280.mm.1 | NM_025526.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010280.mm.1 | NM_025526.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010280.mm.1 | TC0500001210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010279.mm.1 | NM_025526.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010279.mm.1 | NM_025526.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010279.mm.1 | TC0500001210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010278.mm.1 | NM_025526.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010278.mm.1 | NM_025526.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010278.mm.1 | TC0500001210.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452579_at | - | - |
| Mouse Expression Set 430 | 1452579_PM_at | - | - |
| Murine Genome U74v2 Set | 95645_PM_at | - | - |
| Murine Genome U74v2 Set | 95645_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452579_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452579_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452579_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452579_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3060519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234425 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234425 | - | - |
| mouse-6 v1.1 (Illumina) | scl066383.5_131-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066383.5_131-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234425 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234425 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066383.5_131-S | - | - |

Iscu
Genomic Map