Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012027.2 | NP_036157.2 | 11 - 12 | 1606 | 85 | |
| NM_201245.3 | NP_957697.1 | 11 - 12 | 1606 | 85 | ||
| GenBank mRNA | AK035435.1 | - | 5 - 6 | 896 | 85 | |
| AK044512.1 | - | 10 - 11 | 1248 | 85 | ||
| AK154807.1 | - | 12 - 13 | 1613 | 85 | ||
| AK155630.1 | - | 11 - 12 | 1387 | 85 | ||
| AK158296.1 | - | 11 - 12 | 1595 | 85 | ||
| AK162530.1 | - | 9 - 10 | 952 | 85 | ||
| AK170849.1 | - | 11 - 12 | 1590 | 85 | ||
| AK171089.1 | - | 11 - 12 | 1595 | 85 | ||
| BC039932.1 | - | 10 - 11 | 1130 | 85 | ||
| BC049803.1 | - | 11 - 12 | 1132 | 85 | ||
| BC063067.1 | - | 11 - 12 | 1569 | 5721396 | 85 | |
| U73200.1 | - | 11 - 12 | 1466 | 85 | ||
More Information
Set Membership: |
|

Mprip
Genomic Map