Assay Details
| Gene Symbol: Rhoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081105.1 | NP_001074574.1 | 1 - 2 | 314 | 100 | |
| GenBank mRNA | AK017885.1 | - | 1 - 2 | 300 | 100 | |
| AK169618.1 | - | 1 - 2 | 275 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017885.1, AK169618.1, NM_001081105.1 transcripts.
Set Membership: |
|

Rhoh
Genomic Map