Assay Details
| Gene Symbol: Tmprss7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172455.3 | NP_766043.3 | 4 - 5 | 459 | 113 | |
| GenBank mRNA | DQ061860.1 | - | 4 - 5 | 460 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss7
Genomic Map