Assay Details
| Gene Symbol: Ndufa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026614.2 | NP_080890.1 | 4 - 5 | 384 | 96 | |
| GenBank mRNA | AK008155.1 | - | 4 - 5 | 254 | 96 | |
| AK008156.1 | - | 4 - 5 | 250 | 96 | ||
| AK008286.1 | - | 4 - 5 | 250 | 96 | ||
| AK008414.1 | - | 4 - 5 | 250 | 96 | ||
| AK009338.1 | - | 4 - 5 | 268 | 96 | ||
| AK009396.1 | - | 4 - 5 | 254 | 96 | ||
| AK009461.1 | - | 4 - 5 | 254 | 96 | ||
| AK009493.1 | - | 4 - 5 | 253 | 96 | ||
| AK009608.1 | - | 4 - 5 | 254 | 96 | ||
| AK009642.1 | - | 4 - 5 | 269 | 96 | ||
| AK009652.1 | - | 4 - 5 | 254 | 96 | ||
| AK009775.1 | - | 4 - 5 | 278 | 96 | ||
| AK009840.1 | - | 4 - 5 | 268 | 96 | ||
| AK009985.1 | - | 4 - 5 | 268 | 96 | ||
| AK010024.1 | - | 4 - 5 | 253 | 96 | ||
| AK010090.1 | - | 4 - 5 | 256 | 96 | ||
| AK010092.1 | - | 4 - 5 | 270 | 96 | ||
| AK011579.1 | - | 4 - 5 | 244 | 96 | ||
| AK011947.1 | - | 4 - 5 | 244 | 96 | ||
| AK019084.1 | - | 4 - 5 | 255 | 96 | ||
| AK019201.1 | - | 4 - 5 | 255 | 96 | ||
| AK019207.1 | - | 4 - 5 | 246 | 96 | ||
| AK019215.1 | - | 4 - 5 | 254 | 96 | ||
| AK019245.1 | - | 4 - 5 | 275 | 96 | ||
| AK019291.1 | - | 4 - 5 | 255 | 96 | ||
| AK019301.1 | - | 4 - 5 | 385 | 96 | ||
| AK019324.1 | - | 4 - 5 | 364 | 96 | ||
| AK019354.1 | - | 4 - 5 | 257 | 96 | ||
| AK019359.1 | - | 4 - 5 | 276 | 96 | ||
| AK019364.1 | - | 4 - 5 | 270 | 96 | ||
| AK019368.1 | - | 4 - 5 | 268 | 96 | ||
| BC028633.1 | - | 4 - 5 | 252 | 1247576 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302047 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302045 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027698 | AK019291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027698 | AK019301.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027698 | AK019359.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027698 | AK019364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027698 | AK019430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103351 | AK019291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103351 | AK019301.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103351 | AK019324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103351 | AK019354.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103351 | AK019359.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260706 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632116 | AK162445.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42394 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146961 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103351 | AK019291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103351 | AK019301.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103351 | AK019324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103351 | AK019354.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103351 | AK019359.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027698 | AK019291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027698 | AK019301.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027698 | AK019359.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027698 | AK019364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027698 | AK019430.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170156 | AK019291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170156 | AK019301.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170156 | AK019324.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170156 | AK019354.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170156 | AK019359.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24301990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24302046 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146961 | - | - |
| Whole Mouse Genome G4122A | A_51_P170156 | AK019291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170156 | AK019301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170156 | AK019324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170156 | AK019354.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170156 | AK019359.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121725 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05227780 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05227774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05227764 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05227771 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24302047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24302042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24301981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24301998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05864777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24301993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05626029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05227773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05227780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05488501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05227765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05499706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24301989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24301996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05864774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05626037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05499701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24302036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24301994 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10543444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543445 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417286_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417285_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543442 | - | - |
| Mouse Genome 430 2.0 Array | 1417286_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417285_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417285_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417286_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68202_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68202_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600015231.mm.1 | NM_026614.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015231.mm.1 | NM_026614.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015231.mm.1 | TC0600001943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600015232.mm.1 | NM_026614.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015232.mm.1 | NM_026614.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015232.mm.1 | TC0600001943.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417286_PM_at | - | - |
| Mouse Expression Set 430 | 1417285_a_at | - | - |
| Mouse Expression Set 430 | 1417285_PM_a_at | - | - |
| Mouse Expression Set 430 | 1417286_at | - | - |
| Murine Genome U74v2 Set | 97201_PM_s_at | - | - |
| Murine Genome U74v2 Set | 97201_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417286_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417285_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417286_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417285_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417286_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417285_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417286_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417285_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6550035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691460 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216007 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691460 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216007 | - | - |
| mouse-6 v1.1 (Illumina) | scl0068202.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001121.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550035 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001121.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068202.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691460 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1216007 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691460 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216007 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001121.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0068202.1_15-S | - | - |

Ndufa5
Genomic Map