| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1447896_PM_s_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1435524_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10501922 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300022778.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022778.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022778.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022775.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022775.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022775.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022779.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022779.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022779.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022781.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022781.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022781.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022776.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022776.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022776.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022777.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022777.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022777.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022782.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022782.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022782.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300022780.mm.1 |
NR_028573.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022780.mm.1 |
NR_028573.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022780.mm.1 |
NR_028574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022780.mm.1 |
NR_028574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300022780.mm.1 |
TC0300002878.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1435524_at |
- |
- |
| Mouse Expression Set 430 |
1435524_PM_at |
- |
- |
| Mouse Expression Set 430 |
1447896_s_at |
- |
- |
| Mouse Expression Set 430 |
1447896_PM_s_at |
- |
- |
| Murine Genome U74v2 Set |
165863_r_at |
- |
- |
| Murine Genome U74v2 Set |
165862_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
165862_f_at |
- |
- |
| Murine Genome U74v2 Set |
165863_PM_r_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1435524_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1435524_at |
- |
- |