Assay Details
| Gene Symbol: Gm10432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045741.1 | 2 - 3 | 290 | 71 | ||
| GenBank mRNA | AK039577.1 | - | 146 | 71 | ||
| AK147460.1 | - | 2 - 3 | 290 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin 1 | Calm1 |
AI256814, AI327027, AI461935, AL024000, CaM, Calm |
-80.4kb | Sense | Chr. 12 - 100199541 - 100209806 on NCBI Build GRCm38 |
| Downstream | tetratricopeptide repeat domain 7B | Ttc7b |
AA408451, Ttc7l1 |
5.0kb | Anti-sense | Chr. 12 - 100300770 - 100520822 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147460.1 transcripts.
Set Membership: |
|

Gm10432
Genomic Map